Screw Vial Convenience Kit
Screw Vial Convenience Kit
Actual product may vary
Screw Vial Convenience Kit
Store your samples in these vials to eliminate leaching of ions while maintaining a consistent pH for the life of your sample. Storage vials range from 2mL to 40mL.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)ColorClosure Type
B7990-38 mLClearScrew Cap, Open Top
B7990-24 mLClearScrew Cap, Open Top
B7990-1212 mLClearScrew Cap, Open Top
B7990-12A12 mLAmberScrew Cap, Open Top
Catalog number B7990-3
HSN 70179090
Price (MYR)
0.00
Box
Add to cart
Volume (Metric):
8 mL
Color:
Clear
Closure Type:
Screw Cap, Open Top
Price (MYR)
0.00
Box
Add to cart
Store your samples in these vials to eliminate leaching of ions while maintaining a consistent pH for the life of your sample. Thermo Scientific™ National™ and Thermo Scientific™ Chromacol™ Sample Storage Screw Thread Vials are manufactured from superior quality 33 expansion borosilicate clear (Type 1, Class A) or 51A amber (Type 1, Class B) glass. Storage vials range from 2mL to 40mL.
  • PTFE-lined solid-top storage caps
  • PTFE on film/foam backing offers broad chemical resistance
  • Not autoclavable
Order as individual sample storage vials and closures or as unassembled convenience kits.
Unassembled convenience kits saves time during sample preparation.
  • Solid-top convenience kits include matched quantities of vials and screw caps with pre-assembled septa.
  • Open-top convenience kits contain shrink-wrapped vials and separately packaged caps and septa in polybags.
Specifications
DescriptionScrew Vial Convenience Kit, Open top Cap
ColorClear
MaterialGlass
Volume (Metric)8 mL
Includes8mL clear glass vial with open top septa caps
No. per Pack200
Closure TypeScrew Cap, Open Top
Unit SizeBox