2-Nitrophenylselenocyanate, 97%
2-Nitrophenylselenocyanate, 97%
2-Nitrophenylselenocyanate, 97%
2-Nitrophenylselenocyanate, 97%
Thermo Scientific Chemicals
2-Nitrophenylselenocyanate, 97%
CAS: 51694-22-5 | C7H4N2O2Se | 227.092 g/mol
Have Questions?
Catalog NumberQuantity
ALFL20193.031 g
Catalog number ALFL20193.03
Price (MYR)
479.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
479.00
EA
Add to cart
Specifications
Chemical Name or Material2-Nitrophenylselenocyanate
CAS51694-22-5
Health Hazard 1H301+H331-H373
Health Hazard 2GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Health Hazard 3P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c
View more
2-Nitrophenylselenocyanate is used to study the mechanism of its reaction with the zinc/thiolate clusters of metallothionein. It is also used in the preparation of 2,3-seco-5 alfa-cholestane-2,3-diol and in the preparation of 4alfa-methyl-2,3-seco-5 alfa-cholestane-2,3-diol. Further, it is used in the preparation of 2-nitrophenylselenyl derivative and 3alfa-[(2-Nitrophenyl)seleno]androsta-1,5-dien-17beta-ol. In addition to this, it reacts with 6-bromo-hexanoic acid methyl ester to get 6-(2-nitro-phenylselanyl)-hexanoic acid methyl ester.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Nitrophenylselenocyanate is used to study the mechanism of its reaction with the zinc/thiolate clusters of metallothionein. It is also used in the preparation of 2,3-seco-5 alfa-cholestane-2,3-diol and in the preparation of 4alfa-methyl-2,3-seco-5 alfa-cholestane-2,3-diol. Further, it is used in the preparation of 2-nitrophenylselenyl derivative and 3alfa-[(2-Nitrophenyl)seleno]androsta-1,5-dien-17beta-ol. In addition to this, it reacts with 6-bromo-hexanoic acid methyl ester to get 6-(2-nitro-phenylselanyl)-hexanoic acid methyl ester.
Solubility
Soluble in dichloromethane, tetrahydrofuran, dimethyl formamide and ethanol.
Notes
Incompatible with acids and strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Kashinath, K.; Dhara, S.; Reddy, D. S. Breaking and Making of Olefins Simultaneously Using Ozonolysis: Application to the Synthesis of Useful Building Blocks and Macrocyclic Core of Solomonamides. Org. Lett. 2015, 17 (9), 2090-2093.
  2. Bender, M.; Schmidtmann, M.; Summons, R. E.; Rullkötter, J.; Christoffers, J. A Geomimetic Approach to the Formation and Identification of Fossil Sterane Biomarkers in Crude Oil: 18-nor-D-homo-Androstane and 5 alpha,14 beta-Androstane. Chem. Eur. J. 2015, 21 (35), 12501-12508.