Dibenzothiophene-4-boronic acid, 95%
Dibenzothiophene-4-boronic acid, 95%
Dibenzothiophene-4-boronic acid, 95%
Dibenzothiophene-4-boronic acid, 95%
Thermo Scientific Chemicals
Dibenzothiophene-4-boronic acid, 95%
CAS: 108847-20-7 | C12H9BO2S | 228.07 g/mol
Have Questions?
Catalog NumberQuantity
ALFL19831.031 g
Catalog number ALFL19831.03
Price (MYR)
435.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
435.00
EA
Add to cart
Specifications
Chemical Name or MaterialDibenzothiophene-4-boronic acid
CAS108847-20-7
Health Hazard 1H302+H312+H332-H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c