1-n-Butyl-3-methylimidazolium tetrafluoroborate, 98+%
1-n-Butyl-3-methylimidazolium tetrafluoroborate, 98+%
1-n-Butyl-3-methylimidazolium tetrafluoroborate, 98+%
1-n-Butyl-3-methylimidazolium tetrafluoroborate, 98+%
Thermo Scientific Chemicals
1-n-Butyl-3-methylimidazolium tetrafluoroborate, 98+%
CAS: 174501-65-6 | C8H15BF4N2 | 226.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number ALFL19087.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or Material1-n-Butyl-3-methylimidazolium tetrafluoroborate
CAS174501-65-6
Health Hazard 1H301-H315-H319
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
View more
1-n-Butyl-3-methylimidazolium tetrafluoroborate is an ionic liquid used for various reactions such as hydrogenations, asymmetric hydrogenations proceed in higher enantioselectivity than in homogeneous phase. It is also used in Suzuki cross-coupling.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-n-Butyl-3-methylimidazolium tetrafluoroborate is an ionic liquid used for various reactions such as hydrogenations, asymmetric hydrogenations proceed in higher enantioselectivity than in homogeneous phase. It is also used in Suzuki cross-coupling.
Solubility
Miscible with acetone, acetonitrile, ethyl acetate, isopropyl alcohol and methylene chloride. Immiscible with hexane, toluene and water.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Ionic liquid.
  2. Effective medium for fluorination of alkyl halides with KF: J. Am. Chem. Soc., 124, 10278 (2002).
  3. The Suzuki coupling of arylboronic acids with aryl bromides gave improved yields with reduced catalyst concentrations (Tetrakis(triphenyl phosphine) palladium(0) , 10548) with this reaction medium: Chem. Commun., 1249 (2000). Acceleration of Suzuki coupling of polymer-supported 4-iodophenol with arylboronic acids has also been observed: Org. Lett., 4, 3071 (2002).
  4. Akçay, A.; Babucci, M.; Balci, V.; Uzun, A. A model to predict maximum tolerable temperatures of metal-oxide-supported 1-n-butyl-3-methylimidazolium based ionic liquids. Chem. Eng. Sci. 2015, 123, 588-595.
  5. Shahid, R.; Muhammad, N.; Gonfa, G.; Toprak, M. S.; Muhammed, M. Synthesis, COSMO-RS analysis and optical properties of surface modified ZnS quantum dots using ionic liquids. J. Phys. Chem. Solids 2015, 85, 34-38.