n-Hexyltriethoxysilane, 97%
n-Hexyltriethoxysilane, 97%
n-Hexyltriethoxysilane, 97%
n-Hexyltriethoxysilane, 97%
Thermo Scientific Chemicals
n-Hexyltriethoxysilane, 97%
CAS: 18166-37-5 | C12H28O3Si | 248.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFL16521.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Boiling Point115°C (18mmHg)
Chemical Name or Materialn-Hexyltriethoxysilane
CAS18166-37-5
Health Hazard 1H315-H319
Health Hazard 2GHS H Statement
H315-H319