Ethyltriethoxysilane, 96%
Ethyltriethoxysilane, 96%
Ethyltriethoxysilane, 96%
Ethyltriethoxysilane, 96%
Thermo Scientific Chemicals
Ethyltriethoxysilane, 96%
CAS: 78-07-9 | C8H20O3Si | 192.33 g/mol
Have Questions?
Catalog NumberQuantity
ALFL16495.AC25 mL
Catalog number ALFL16495.AC
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 mL
Request bulk or custom format
Specifications
Boiling Point158°C to 159°C
Chemical Name or MaterialEthyltriethoxysilane
Melting Point-78°C
CAS78-07-9
Health Hazard 1H226