Tri-n-octylphosphine, tech. 90%
Tri-n-octylphosphine, tech. 90%
Tri-n-octylphosphine, tech. 90%
Tri-n-octylphosphine, tech. 90%
Thermo Scientific Chemicals
Tri-n-octylphosphine, tech. 90%
CAS: 4731-53-7 | C24H51P | 370.646 g/mol
Have Questions?
Catalog NumberQuantity
ALFL15744.AC25 mL
Catalog number ALFL15744.AC
Price (MYR)
514.00
EA
Add to cart
Quantity:
25 mL
Request bulk or custom format
Price (MYR)
514.00
EA
Add to cart
Specifications
Chemical Name or MaterialTri-n-octylphosphine
CAS4731-53-7
Health Hazard 1H314-H335
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
View more
Tri-n-octylphosphine is used for coating zinc sulfide shells on cadmium-selenium quantum dot core by successive ionic layer adsorption and reaction method. It acts as a precursor to trioctylphosphine oxide. Further, it is used as a solvent for cadmium and selenium precursors. It also serves as a common reagent in the chemical synthesis of nanoparticles. It acts as a precursor to trioctylphosphine oxide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tri-n-octylphosphine is used for coating zinc sulfide shells on cadmium-selenium quantum dot core by successive ionic layer adsorption and reaction method. It acts as a precursor to trioctylphosphine oxide. Further, it is used as a solvent for cadmium and selenium precursors. It also serves as a common reagent in the chemical synthesis of nanoparticles. It acts as a precursor to trioctylphosphine oxide.
Solubility
Immiscible with water.
Notes
Air sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Mntungwa, N.; Khan, M. D.; Mlowe, S.; Revaprasadu, N. A simple route to alkylamine capped antimony nanoparticles. Mater. Lett. 2015, 145, 239-242.
  2. More, D. S.; Moloto, M. J.; Moloto, N.; Matabola, K. P. TOPO-capped silver selenide nanoparticles and their incorporation into polymer nanofibers using electrospinning technique. Mater. Res. Bull. 2015, 65, 14-22.