4,4'-Oxybis(benzoic acid), 98+%
4,4'-Oxybis(benzoic acid), 98+%
4,4'-Oxybis(benzoic acid), 98+%
4,4'-Oxybis(benzoic acid), 98+%
Thermo Scientific Chemicals
4,4'-Oxybis(benzoic acid), 98+%
CAS: 2215-89-6 | C14H10O5 | 258.229 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALFL15651.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material4,4'-Oxybis(benzoic acid)
CAS2215-89-6
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c