Eaton's Reagent, P{2}O{5} ca 7.5% w/w
Eaton's Reagent, P{2}O{5} ca 7.5% w/w
Eaton's Reagent, P{2}O{5} ca 7.5% w/w
Eaton's Reagent, P{2}O{5} ca 7.5% w/w
Thermo Scientific Chemicals
Eaton's Reagent, P{2}O{5} ca 7.5% w/w
CAS: 39394-84-8 | CH4O8P2S | 238.043 g/mol
Have Questions?
Catalog NumberQuantity
ALFL15607.AE100 mL
Catalog number ALFL15607.AE
Price (MYR)
408.00
EA
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
408.00
EA
Add to cart
Specifications
Chemical Name or MaterialEaton's Reagent
Name NoteP2O5
CAS39394-84-8
Health Hazard 1H290-H302+H312-H314-H335
Health Hazard 2GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
View more
Eaton's Reagent is used in the synthesis fluorine-based hole-transporting material, mono and bis-chalcone derivatives via Claisen-Schmidt condensation reaction between arylaldehydes and ketones and 4-hydroxycoumarins and 4-hydroxy-2-quinolinones derivatives.