Amberlite™ IRC120 Na ion exchange resin
Amberlite™ IRC120 Na ion exchange resin
Amberlite™ IRC120 Na ion exchange resin
Amberlite™ IRC120 Na ion exchange resin
Thermo Scientific Chemicals
Amberlite™ IRC120 Na ion exchange resin
CAS: 78922-04-0 | C13H10ClNO4S | 311.736 g/mol
Have Questions?
Catalog NumberQuantity
ALFL15199.36500 g
Catalog number ALFL15199.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAmberlite™ IR-120(Na) ion exchange resin
CAS78922-04-0
Recommended StorageAmbient temperatures
EINECS Number000-000-0
TSCANo
View more
Amberlite™ IR-120(Na) ion exchange resin is used in water softening, dealkalinization and deionization. It is used for the recovery of toxic metals. It is also used in separation medium for TLC, paper chromatography and LPLC.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Amberlite™ IR-120(Na) ion exchange resin is used in water softening, dealkalinization and deionization. It is used for the recovery of toxic metals. It is also used in separation medium for TLC, paper chromatography and LPLC.
Solubility
Insoluble in water.
Notes
Water softening, dealkalinization, deionization
RUO – Research Use Only
General References:
  1. José L. Valverde.; Antonio de Lucas.; Marcela González.; Juan F. Rodríguez. Ion-Exchange Equilibria of Cu2+, Cd2+, Zn2+, and Na+ Ions on the Cationic Exchanger Amberlite IR-120. J. Chem. Eng. Data. 2001, 46 1404-1409.
  2. Juan Francisco Rodríguez.; Antonio De Lucas.; Jose R. Leal.; Jose Luis Valverde. Determination of intraparticle diffusivities of Na+/K+ in water and water/alcohol mixed solvents on a strong acid cation exchanger. Ind. Eng. Chem. Res. 2002, 41 3019-3027.