Adenosine-5'-triphosphate disodium salt hydrate, 99%, water <10%
Adenosine-5'-triphosphate disodium salt hydrate, 99%, water &lt;10%
Adenosine-5'-triphosphate disodium salt hydrate, 99%, water &lt;10%
Adenosine-5'-triphosphate disodium salt hydrate, 99%, water &lt;10%
Thermo Scientific Chemicals
Adenosine-5'-triphosphate disodium salt hydrate, 99%, water <10%
CAS: 34369-07-8 | C10H14N5Na2O13P3 | 551.15 g/mol
Have Questions?
Catalog NumberQuantity
ALFL14522.031 g
Catalog number ALFL14522.03
Price (MYR)
480.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
480.00
EA
Add to cart
Specifications
Chemical Name or MaterialAdenosine-5'-triphosphate disodium salt hydrate
CAS34369-07-8
Recommended StorageStore at -20°C
RTECS NumberAU7417000
EINECS Number213-579-1
View more
Adenosine-5'-triphosphate disodium salt hydrate is used as a form of cellular energy. It is used in many cellular processes, respiration, biosynthetic reactions, motility, and cell division. ATP is a substrate of many kinases involved in cell signaling and of adenylate cyclase(s) that produce the second messenger cAMP. ATP provides the metabolic energy to drive metabolic pumps. ATP serves as a coenzyme in a wide array of enzymatic reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Adenosine-5′-triphosphate disodium salt hydrate is used as a form of cellular energy. It is used in many cellular processes, respiration, biosynthetic reactions, motility, and cell division. ATP is a substrate of many kinases involved in cell signaling and of adenylate cyclase(s) that produce the second messenger cAMP. ATP provides the metabolic energy to drive metabolic pumps. ATP serves as a coenzyme in a wide array of enzymatic reactions.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is -20°C. Store under inert gas. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. Chao Qi.; Qi-Li Tang.; Ying-Jie Zhu.; Xin-Yu Zhao.; Feng Chen. Microwave-assisted hydrothermal rapid synthesis of hydroxyapatite nanowires using adenosine 5'-triphosphate disodium salt as phosphorus source. 2012, 85 71-73.
  2. Emil Pop.; Viorel Soran.; Georgeta Lazǎr. The Effect of ATP (Disodium Salt) upon Rotational Streaming. Physiologia Plantarum. 1967, 20 (3), 617-623.