Methacrylic anhydride, 94%, stab. with ca 0.2% 2,4-dimethyl-6-tert-butylphenol
Methacrylic anhydride, 94%, stab. with ca 0.2% 2,4-dimethyl-6-tert-butylphenol
Methacrylic anhydride, 94%, stab. with ca 0.2% 2,4-dimethyl-6-tert-butylphenol
Methacrylic anhydride, 94%, stab. with ca 0.2% 2,4-dimethyl-6-tert-butylphenol
Thermo Scientific Chemicals
Methacrylic anhydride, 94%, stab. with ca 0.2% 2,4-dimethyl-6-tert-butylphenol
CAS: 760-93-0 | C8H10O3 | 154.17 g/mol
Have Questions?
Catalog NumberQuantity
ALFL14357.1850 g
Catalog number ALFL14357.18
Price (MYR)
352.00
EA
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (MYR)
352.00
EA
Add to cart
Specifications
Chemical Name or MaterialMethacrylic anhydride
Name Notestabilized with ≈0.2% 2,4-dimethyl-6-tert-butylphenol
CAS760-93-0
Health Hazard 1H227-H302+H332-H315-H317-H318-H335
Health Hazard 2GHS H Statement
H330-H314-H318-H227
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
View more
Methacrylic anhydride is a reactive monomer used for the preparation of different type of monomers under mild reaction conditions. It finds applications in light-curing coatings, cross-linked materials like synthetic resin. It acts as an intermediate for the preparation of fibers and resins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methacrylic anhydride is a reactive monomer used for the preparation of different type of monomers under mild reaction conditions. It finds applications in light-curing coatings, cross-linked materials like synthetic resin. It acts as an intermediate for the preparation of fibers and resins.
Solubility
Miscible with ethanol and ether.
Notes
Moisture and light sensitive. Store in a cool place. Incompatible with acids, strong oxidizing agents, alcohols, amines and bases.
RUO – Research Use Only
General References:
  1. Erwin, Z.; Blokzijl, M. M.; Grijpma, D. W. A Combinatorial Photocrosslinking Method for the Preparation of Porous Structures with Widely Differing Properties. Macromol. Rapid Commun. 2015, 36 (21), 1902-1909.
  2. Li, M.; Hu, Y.; Li, Y.; Liu, Y.; Zeng, C.; Liu, Q.; Hou, C. Synthesis and Characterization of Polymerizable Tourmaline Methacrylate. e-J. Surf. Sci. Nanotechnol. 2015, 13, 422-425.