Tetrafluoroboric acid, ca 50% w/w aq. soln.
Tetrafluoroboric acid, ca 50% w/w aq. soln.
Tetrafluoroboric acid, ca 50% w/w aq. soln.
Tetrafluoroboric acid, ca 50% w/w aq. soln.
Thermo Scientific Chemicals
Tetrafluoroboric acid, ca 50% w/w aq. soln.
CAS: 16872-11-0
Have Questions?
Catalog NumberQuantity
ALFL14037.36500 g
Catalog number ALFL14037.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialTetrafluoroboric acid
CAS16872-11-0
Boiling Point130°C (decomposition)
Concentration≈50% w/w aq. soln.
Health Hazard 1H314-H360FD
View more
Tetrafluoroboric acid is used in conjunction with various methods of cleaning and to treat the surface of substrates. It is also used as a radiotracer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tetrafluoroboric acid is used in conjunction with various methods of cleaning and to treat the surface of substrates. It is also used as a radiotracer.
Solubility
Miscible with water.
Notes
Incompatible with strong bases, oxidizing agents and acids.
RUO – Research Use Only
General References:
  1. Precursor of tetrafluoroborate salts of various stabilized cations. See, e.g.: Org. Synth. Coll., 5, 1135 (pyrylium), 1138 (tropylium); 6, 364 (sulfonium), 991 (cyclopropenium).
  2. Addition to aqueous solutions of aryldiazonium salts usually results in precipitation of the diazonium tetrafluoroborates, which are often stable enough to be isolated, and can undergo various reactions in non-aqueous solvents. For hydrodediazoniation in the presence of formamide, see: J. Chem. Soc., Perkin 1, 873 (1986). Probably their most important use is the Schiemann (or Balz-Schiemann) synthesis of aryl fluorides in which the tetrafluoroborate salt is heated neat or in an inert solvent. For examples, see: Org. Synth. Coll., 2, 188, 295, 299 (1943). Reviews: Org. React., 5, 193 (1949); Adv. Fluorine. Chem., 4, 1 (1965). See also Hexafluorophosphoric acid, L15728.
  3. Chen, L.; Cao, X.; Huang, D.; Pan, Q. Switchable dielectric phase transition in tris(1-(chloromethyl)-1,4-diazabicyclo[2.2.2]octane) tetra(tetrafluoroborate) dichloride. Inorg. Chem. Commun. 2015, 61, 93-96.
  4. Zhong, Z.; Brokmeiera, H.; Maawad, E.; Schell, N. In-situ investigation of the anisotropic mechanical behavior of rolled AA 7020-T6 alloy through lattice strain evolution during uniaxial tension. Mater. Sci. Eng., A 2015, 639, 519-525.