Perfluorotetradecanoic acid, 96%
Perfluorotetradecanoic acid, 96%
Perfluorotetradecanoic acid, 96%
Perfluorotetradecanoic acid, 96%
Thermo Scientific Chemicals
Perfluorotetradecanoic acid, 96%
CAS: 376-06-7 | C14HF27O2 | 714.117 g/mol
Have Questions?
Catalog NumberQuantity
ALFL13796.031 g
Catalog number ALFL13796.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPerfluorotetradecanoic acid
CAS376-06-7
Health Hazard 1H314-H335
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
View more
Perfluorotetradecanoic acid is used for GC (Gas Chromatography) and LC (Liquid Chromatography) analysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Perfluorotetradecanoic acid is used for GC (Gas Chromatography) and LC (Liquid Chromatography) analysis.
Solubility
It is insoluble in water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only
General References:
  1. HJ Lehmler.; PM Bummer. Mixing of perfluorinated carboxylic acids with dipalmitoylphosphatidylcholine. Biochimica et Biophysica Acta . 2004, 1664,(2), 141-149.
  2. HK Park.; TH Ha.; K Kim. Gelation of octadecyltrimethoxysilane at the air/water interface: Effect ofperfluorotetradecanoic acid and divalent cadmium ions. Langmuir. 2004, 20,(12), 4851-4858.