Petroleum ether 60/80
Petroleum ether 60/80
Thermo Scientific Chemicals
Petroleum ether 60/80
CAS: 8032-32-4
Have Questions?
Change viewbuttonViewtableView
Quantity:
1000 mL
2500 mL
Catalog number ALFL13734.AU
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1000 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialPetroleum ether 60/80
CAS8032-32-4
Health Hazard 1H225-H304-H315-H319-H335-H336
Health Hazard 2GHS H Statement
H225-H340-H350-H304
Highly flammable liquid and vapor.
May cause genetic defects.
May cause cancer.
May be fatal if swallowed and enters airways.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
View more
It is mostly used by pharmaceutical companies and in the manufacturing process. It is a lightweight hydrocarbon used chiefly as a nonpolar solvent. The extract of aerial parts of ageratum conyzoids in petroleum ether plays an essential role for the isolation of sigmasterol and beta sitosterol. Crude petroleum spirit and methanol extracts also play an important role for the screening of antimicrobial activity of the stem bark, leaves and roots of Ficus thoningii.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is mostly used by pharmaceutical companies and in the manufacturing process. It is a lightweight hydrocarbon used chiefly as a nonpolar solvent. The extract of aerial parts of ageratum conyzoids in petroleum ether plays an essential role for the isolation of sigmasterol and beta sitosterol. Crude petroleum spirit and methanol extracts also play an important role for the screening of antimicrobial activity of the stem bark, leaves and roots of Ficus thoningii.
Solubility
Insoluble in water.
Notes
Flammable, may be moisture sensitive.
RUO – Research Use Only
General References:
  1. Kamboj, A.; Saluja, A. K. Isolation of Stigmasterol and beta sitosterol from petroleum ether extract of Aerial parts of Ageratum Conyzoides (Asteracea). Inter. J. Pharma. Pharma. Sci. 2011, 3 (1), 94­96.
  2. Ndukwe, I. G.; Bello, A. I.; Habila, J. D.; John, C. Phytochemical and antimicrobial screening of the crude petroleum spirit and methanol extracts of the stem bark, leaves and roots of Ficus thoningii (blume). Afr. J. Biotechnol. 2007, 6 (23), 2645-2649.