2-Nitroimidazole, 98%
2-Nitroimidazole, 98%
2-Nitroimidazole, 98%
2-Nitroimidazole, 98%
Thermo Scientific Chemicals
2-Nitroimidazole, 98%
CAS: 527-73-1 | C3H3N3O2 | 113.08 g/mol
Have Questions?
Catalog NumberQuantity
ALFL13546.MC100 mg
Catalog number ALFL13546.MC
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mg
Request bulk or custom format
Specifications
Chemical Name or Material2-Nitroimidazole
CAS527-73-1
Health Hazard 1H301-H315-H319-H335
Health Hazard 2GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
View more
2-Nitroimidazole, is used as an antibiotic in pharma industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Nitroimidazole, is used as an antibiotic in pharma industry.
Solubility
It is slightly soluble in water. Soluble in acetone, butyl acetate, ethanol, ethyl acetate and methanol.
Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents, strong acids, acid chlorides, anhydrous acids.. Stable under recommended storage conditions.
RUO – Research Use Only
General References:
  1. EM Lord.; L Harwel.; CJ Koch. Detection of hypoxic cells by monoclonal antibody recognizing 2-nitroimidazole adducts. Cancer research. 1993, 53,(1), 5721-5731.
  2. BA Moore.; B Palcic.; LD Skarsgard. Radiosensitizing and toxic effects of the 2-nitroimidazole Ro-07-0582 in hypoxic mammalian cells. Radiation research. 1976, 67,(3), 459-473.