2-(Dimethylamino)ethyl methacrylate, 97%, stab. with ca 0.2% 4-methoxyphenol
2-(Dimethylamino)ethyl methacrylate, 97%, stab. with ca 0.2% 4-methoxyphenol
2-(Dimethylamino)ethyl methacrylate, 97%, stab. with ca 0.2% 4-methoxyphenol
2-(Dimethylamino)ethyl methacrylate, 97%, stab. with ca 0.2% 4-methoxyphenol
Thermo Scientific Chemicals
2-(Dimethylamino)ethyl methacrylate, 97%, stab. with ca 0.2% 4-methoxyphenol
CAS: 2867-47-2 | C8H15NO2 | 157.213 g/mol
Have Questions?
Catalog NumberQuantity
ALFL13507.AE100 mL
Catalog number ALFL13507.AE
Price (MYR)
355.00
EA
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
355.00
EA
Add to cart
Specifications
Chemical Name or Material2-(Dimethylamino)ethyl methacrylate
Name Notestabilized with ≈0.2% 4-methoxyphenol
CAS2867-47-2
Health Hazard 1H227-H302-H315-H317-H319-H330
Health Hazard 2GHS H Statement
H302-H312-H315-H319-H317-H227
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Combustible liquid.
View more
2-(Dimethylamino)ethyl methacrylate is used in the synthesis of poly ((2-(Dimethylamino)ethyl methacrylate), block copolymers of 2-(Dimethylamino)ethyl methacrylate and n-butyl methacrylate and amphiphilic AB block copolymers of 2-(Dimethylamino)ethyl methacrylate. It is also used in the preparation of polymer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-(Dimethylamino)ethyl methacrylate is used in the synthesis of poly ((2-(Dimethylamino)ethyl methacrylate), block copolymers of 2-(Dimethylamino)ethyl methacrylate and n-butyl methacrylate and amphiphilic AB block copolymers of 2-(Dimethylamino)ethyl methacrylate. It is also used in the preparation of polymer.
Solubility
Soluble in water. (1.000 g/L at 20°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C. It is sensitive to moisture and light. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
  1. J Y Cherng.; P van de Wetering.; H Talsma.; D J Crommelin.; W E Hennink. Freeze-drying of poly((2-dimethylamino)ethyl methacrylate)-based gene delivery systems. Pharmaceutical Research. 1997, 14 (12), 1838-1841.
  2. Sang Beom Lee.; Alan J Russell.; Krzysztof Matyjaszewski. ATRP synthesis of amphiphilic random, gradient, and block copolymers of 2-(dimethylamino)ethyl methacrylate and n-butyl methacrylate in aqueous media. Biomacromolecules. 2003, 4 (5), 1386-1393.