Diethylene glycol monoethyl ether acetate, 99%
Diethylene glycol monoethyl ether acetate, 99%
Diethylene glycol monoethyl ether acetate, 99%
Diethylene glycol monoethyl ether acetate, 99%
Thermo Scientific Chemicals
Diethylene glycol monoethyl ether acetate, 99%
CAS: 112-15-2 | C8H16O4 | 176.21 g/mol
Have Questions?
Catalog NumberQuantity
ALFL13446.AE100 mL
Catalog number ALFL13446.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialDiethylene glycol monoethyl ether acetate
CAS112-15-2
Health Hazard 1H319-H335
Health Hazard 2GHS H Statement
H319
Causes serious eye irritation.
Health Hazard 3P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c
View more
Diethylene glycol monoethyl ether acetate is used as a catalytic agent, petrochemical additive, coating auxiliary agent and in organic synthesis. It is mainly used as metal, furniture paint solvents and as organic intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Diethylene glycol monoethyl ether acetate is used as a catalytic agent, petrochemical additive, coating auxiliary agent and in organic synthesis. It is mainly used as metal, furniture paint solvents and as organic intermediate.
Solubility
Soluble in water.
Notes
Store at room temperature. Incompatible with bases and oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Containers which are opened must be carefully resealed and kept upright to prevent leakage.
RUO – Research Use Only
General References:
  1. R L Smith. Review of glycol ether and glycol ether ester solvents used in the coating industry. Environ Health Perspect. 1984, 57 1-4.
  2. Richard M. Stephenson. Mutual solubilities: water-glycol ethers and water-glycol esters . J. Chem. Eng. Data. 1993, 38 (1), 134-138.