Xylenes, mixed, 97+%
Xylenes, mixed, 97+%
Xylenes, mixed, 97+%
Xylenes, mixed, 97+%
Thermo Scientific Chemicals
Xylenes, mixed, 97+%
CAS: 1330-20-7 | C8H10
Have Questions?
Catalog NumberQuantity
ALFL13317.AP500 mL
Catalog number ALFL13317.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialXylenes
Melting Point<-25°C
Name Notemixed
CAS1330-20-7
Health Hazard 1H226-H304-H312+H332-H315-H319-H335-H373
View more
Xylene is used as a solvent in pharmaceutical, printing, rubber and leather industries. It is an important component of ink, rubber and adhesives as well as thinning paints and varnishes. It is used in the laboratory to make baths with dry ice to cool reaction vessels and as a solvent to remove synthetic immersion oil from the microscope objective in light microscopy. It finds application as a short-term hydrophobic clearing agent of tissue samples by histologists. It is a precursor for the preparation of xylene dichlorides, xylyl bromide, terephthalic acid and dimethyl terephthalate. It acts as a cleaning agent for steel, silicon wafers and integrated circuits.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Xylene is used as a solvent in pharmaceutical, printing, rubber and leather industries. It is an important component of ink, rubber and adhesives as well as thinning paints and varnishes. It is used in the laboratory to make baths with dry ice to cool reaction vessels and as a solvent to remove synthetic immersion oil from the microscope objective in light microscopy. It finds application as a short-term hydrophobic clearing agent of tissue samples by histologists. It is a precursor for the preparation of xylene dichlorides, xylyl bromide, terephthalic acid and dimethyl terephthalate. It acts as a cleaning agent for steel, silicon wafers and integrated circuits.
Solubility
Miscible with aromatic hydrocarbons. Immiscible with water.
Notes
Contains a mixture of the ortho, meta, and para isomers of xylene. May also contain ethylbenzene. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Ferrari-Lima, A. M.; De Souza, R. P.; Mendes, S. S.; Marques, R. G.; Gimenes, M. L.; Machado, N. R. C. F. Photodegradation of benzene, toluene and xylenes under visible light applying N-doped mixed TiO2 and ZnO catalysts. Catal. Today 2015, 241, 40-46.
  2. Nassimbeni, L. R.; Bathori, N. B.; Patel, L. D.; Su, H.; Weber, E. Separation of xylenes by enclathration. Chem. Commun. 2015, 51 (17), 3627-3629.