Hydrogen peroxide, 27% w/w aq. soln., stab.
Hydrogen peroxide, 27% w/w aq. soln., stab.
Hydrogen peroxide, 27% w/w aq. soln., stab.
Hydrogen peroxide, 27% w/w aq. soln., stab.
Thermo Scientific Chemicals
Hydrogen peroxide, 27% w/w aq. soln., stab.
CAS: 7722-84-1 | H2O2 | 34.01 g/mol
Have Questions?
Catalog NumberQuantity
ALFL13235.AE100 mL
Catalog number ALFL13235.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialHydrogen peroxide
Name Notestabilized
CAS7722-84-1
Boiling Point107°C
Health Hazard 1H271-H302+H332-H314
View more
Hydrogen peroxide is used as an oxidizing agent to prepare propylene oxide, dibenzoyl peroxide, lauryl peroxide, peracetic acid and meta-chloroperoxybenzoic acid. It is also involved in the production of inorganic chemicals like sodium perborate and sodium percarbonate, which are used as bleaching agents in detergents. It is an associated iron catalyst and utilized for waste-water treatment to remove organic impurities through an advanced oxidation process. It is also used as a laboratory disinfectant and as a topical antiseptic. In the cosmetic industry, it is used for bleaching human hair and tooth whitening by mixing with ammonium hydroxide and baking soda respectively. Monopropellant engines using hydrogen peroxide as the fuel are commonly used for air-independent underwater propulsion applications.
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
RUO – Research Use Only