Hydrochloric acid, 36% w/w aq. soln.
Hydrochloric acid, 36% w/w aq. soln.
Hydrochloric acid, 36% w/w aq. soln.
Hydrochloric acid, 36% w/w aq. soln.
Thermo Scientific Chemicals
Hydrochloric acid, 36% w/w aq. soln.
CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1000 mL
2500 mL
Catalog number ALFL13091.AU
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1000 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialHydrochloric acid
CAS7647-01-0
Boiling Point81.5°C to 110°C
Concentration36% w/w aq. soln.
Health Hazard 1H290-H314-H335
View more
Hydrochloric acid is used in chemical synthesis reactions to produce vinyl chloride, fertilizers, dyes, pigments and pharmaceutical ingredients. It is also used in catalyst regeneration, rubber manufacturing, electroplating, semiconductor manufacturing, leather tanning and cotton seed processing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Hydrochloric acid is used in chemical synthesis reactions to produce vinyl chloride, fertilizers, dyes, pigments and pharmaceutical ingredients. It is also used in catalyst regeneration, rubber manufacturing, electroplating, semiconductor manufacturing, leather tanning and cotton seed processing.
Solubility
Completely miscible with water.
Notes
Store in cool dry place, away from acids and alkalis.
RUO – Research Use Only
General References:
  1. Jay A. Young. Hydrochloric Acid (approx 36%) Chem. Educ. 2001, 78 (7), 873.
  2. Cui, C.; Hu, Q.; Ren, J.; Zhao, H.; You, L.; Zhao, M. Effect of the Structural Features of Hydrochloric Acid-Deamidated Wheat Gluten on Its Susceptibility to Enzymatic Hydrolysis. Agric. Food Chem. 2013, 61 (24), 5706-5714.