Dichloromethane, 99+%, stab. with ca. 50ppm amylene
Dichloromethane, 99+%, stab. with ca. 50ppm amylene
Dichloromethane, 99+%, stab. with ca. 50ppm amylene
Dichloromethane, 99+%, stab. with ca. 50ppm amylene
Thermo Scientific Chemicals
Dichloromethane, 99+%, stab. with ca. 50ppm amylene
CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol
Have Questions?
Catalog NumberQuantity
ALFL13089.AP500 mL
Catalog number ALFL13089.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialDichloromethane
Melting Point-95°C
Name Notestabilized with ca. 50ppm amylene
CAS75-09-2
Health Hazard 1H315-H319-H336-H350-H373
View more
Dichloromethane is employed in pharmaceutical applications. Its utilization is as a solvent in manufacturing of fine chemicals, drugs, drug intermediates and pharmaceuticals, and also in food engineering. It is the solvent of choice for caffeine extraction. It is employed as a solvent and paint remover, and as a blowing agent for polyurethane foams. It is useful for the fraction of flash pyrolysis oil and for the extraction of benzo-15-crown-5.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Dichloromethane is employed in pharmaceutical applications. Its utilization is as a solvent in manufacturing of fine chemicals, drugs, drug intermediates and pharmaceuticals, and also in food engineering. It is the solvent of choice for caffeine extraction. It is employed as a solvent and paint remover, and as a blowing agent for polyurethane foams. It is useful for the fraction of flash pyrolysis oil and for the extraction of benzo-15-crown-5.
Solubility
Miscible with ethyl acetate, alcohol, hexanes, methanol, diethyl ether, n-octanol, acetone benzene, carbon tetrachloride, diethyl ether and chloroform. Immiscible with water.
Notes
Keep the container tightly closed in a cool, well-ventilated area. Incompatible with alkali metals, aluminum, strong oxidizing agents, bases, amines, magnesium, strong acids strong bases and vinyl compounds.
RUO – Research Use Only
General References:
  1. Berrichi, A.; Bachir, R.; Benabdallah, M.; Choukchou-Braham, N. Supported nano gold catalyzed three-component coupling reactions of amines, dichloromethane and terminal alkynes (AHA). Tetrahedron Lett. 2015, 56 (11), 1302-1306.
  2. Kane, A.; Giraudet, S.; Vilmain, J. B.; Cloirec, P. L. Intensification of the temperature-swing adsorption process with a heat pump for the recovery of dichloromethane. J. Environ. Chem. Eng. 2015, 3 (2), 734-743.