Palmitic acid hydrazide, 98+%
Palmitic acid hydrazide, 98+%
Palmitic acid hydrazide, 98+%
Palmitic acid hydrazide, 98+%
Palmitic acid hydrazide, 98+%
CAS: 2619-88-7 | C16H34N2O | 270.461 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALFL13013.06
Price (MYR)
-
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPalmitic acid hydrazide
CAS2619-88-7
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c