1-Dodecylamine hydrochloride, 97%
1-Dodecylamine hydrochloride, 97%
1-Dodecylamine hydrochloride, 97%
1-Dodecylamine hydrochloride, 97%
Thermo Scientific Chemicals
1-Dodecylamine hydrochloride, 97%
CAS: 929-73-7 | C12H28ClN | 221.81 g/mol
Have Questions?
Catalog NumberQuantity
ALFL12314.1850 g
Catalog number ALFL12314.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or Material1-Dodecylamine hydrochloride
CAS929-73-7
Health Hazard 1H302-H315-H319-H335
Health Hazard 2GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c