2,2-Dimethyl-3-pentanol, 97%, Thermo Scientific™
2,2-Dimethyl-3-pentanol, 97%, Thermo Scientific™
2,2-Dimethyl-3-pentanol, 97%, Thermo Scientific™
2,2-Dimethyl-3-pentanol, 97%, Thermo Scientific™
2,2-Dimethyl-3-pentanol, 97%, Thermo Scientific™
Have Questions?
Catalog NumberQuantity
ALFL11545.031 g
Catalog number ALFL11545.03
Price (MYR)
-
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or Material2,2-Dimethyl-3-pentanol
CAS3970-62-5
Health Hazard 1H226
Health Hazard 2GHS H Statement
H226
Flammable liquid and vapor.
Health Hazard 3P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
View more
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.