4-Nitrophenyl palmitate, 98+%
4-Nitrophenyl palmitate, 98+%
4-Nitrophenyl palmitate, 98+%
4-Nitrophenyl palmitate, 98+%
Thermo Scientific Chemicals
4-Nitrophenyl palmitate, 98+%
CAS: 1492-30-4 | C22H35NO4 | 377.525 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALFL10896.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Nitrophenyl palmitate
CAS1492-30-4
Melting Point65°C to 66°C
Recommended StorageAmbient temperatures
EINECS Number216-084-9
View more
The activities of free and immobilized PPL were determined by spectrophotometric assay using 4-nitrophenyl palmitate (pNPP) as substrate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
The activities of free and immobilized PPL were determined by spectrophotometric assay using 4-nitrophenyl palmitate (pNPP) as substrate.
Solubility
Insoluble in water. Soluble in CHCl3.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Store at -20°C
RUO – Research Use Only
General References:
  1. E. Ranjbakhsh,; A.K. Bordbar,; M. Abbasia,; A.R. Khosropoura,; E. Shams. Supuran. Enhancement of stability and catalytic activity of immobilized lipase on silica-coated modified magnetite nanoparticles. Chemical Engineering Journal. 2012, 179 272-276.
  2. C K Tu, H G Thomas,; G C Wynns,; D N Silverman. Hydrolysis of 4-nitrophenyl acetate catalyzed by carbonic anhydrase III from bovine skeletal muscle. The Journal of Biological Chemistry, . 1986 , 261 10100-10103..