4-(Methoxycarbonyl)phenyl isothiocyanate, 98+%
4-(Methoxycarbonyl)phenyl isothiocyanate, 98+%
4-(Methoxycarbonyl)phenyl isothiocyanate, 98+%
4-(Methoxycarbonyl)phenyl isothiocyanate, 98+%
Thermo Scientific Chemicals
4-(Methoxycarbonyl)phenyl isothiocyanate, 98+%
CAS: 3662-78-0 | C9H7NO2S | 193.22 g/mol
Have Questions?
Catalog NumberQuantity
ALFL10474.065 g
Catalog number ALFL10474.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material4-(Methoxycarbonyl)phenyl isothiocyanate
CAS3662-78-0
Health Hazard 1H302+H312-H314-H331-H335
Health Hazard 2GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c