Dimethyl disulfide, 99%
Dimethyl disulfide, 99%
Dimethyl disulfide, 99%
Dimethyl disulfide, 99%
Thermo Scientific Chemicals
Dimethyl disulfide, 99%
CAS: 624-92-0 | C2H6S2 | 94.19 g/mol
Have Questions?
Catalog NumberQuantity
ALFL09679.30250 g
Catalog number ALFL09679.30
Price (MYR)
348.00
EA
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
348.00
EA
Add to cart
Specifications
Chemical Name or MaterialDimethyl disulfide
CAS624-92-0
Health Hazard 1H225-H301+H331-H317-H319-H335
Health Hazard 2GHS H Statement
H225-H331-H302-H319-H317-H335
Highly flammable liquid and vapor.
Toxic if inhaled.
Harmful if swallowed.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P272-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P311-P330-P333+P313-P363-P370+P378q-P501c
View more
Dimethyl disulfide is used as an intermediate as well as a food additive in onion, garlic, cheese, meats, soups, savory flavors and fruit flavors. It is used in oil refineries as a sulfiding agent. It is used to alfa substitute 2-methylfuran-acrolein to produce food stuff. It is involved in the preparation of 4-(methylthio)phenol. Further, it is used to replace methyl mercaptan as a jet fuel additive. It finds application in artificial flavoring agent and a corrosion inhibitor. In addition to this, it serves as an effective soil fumigant in agriculture.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Dimethyl disulfide is used as an intermediate as well as a food additive in onion, garlic, cheese, meats, soups, savory flavors and fruit flavors. It is used in oil refineries as a sulfiding agent. It is used to alfa substitute 2-methylfuran-acrolein to produce food stuff. It is involved in the preparation of 4-(methylthio)phenol. Further, it is used to replace methyl mercaptan as a jet fuel additive. It finds application in artificial flavoring agent and a corrosion inhibitor. In addition to this, it serves as an effective soil fumigant in agriculture.
Solubility
Miscible with ethanol, ethyl ether, alcohol, most organic solvents and oils. Slightly miscible with water.
Notes
Incompatible with strong bases, strong oxidizing agents and strong reducing agents.
RUO – Research Use Only
General References:
  1. Non-aqueous diazotization of arylamines with isoamyl nitrite in the presence of Me2S2 provides a convenient route to aryl methyl sulfides: Synth. Commun., 31, 1857 (2001). Alkyl methyl sulfides have been prepared by disproportionation of symmetrical disulfides with Me2S2 in the presence of NaOH: Chem. Lett., 755 (1999).
  2. Arellano-García, L.; Dorado, A. D.; Morales-Guadarrama, A.; Sacristan, E.; Gamisans, X.; Revah, S. Modeling the effects of biomass accumulation on the performance of a biotrickling filter packed with PUF support for the alkaline biotreatment of dimethyl disulfide vapors in air. Appl. Microbiol. Biotechnol. 2015, 99 (1), 97-107.
  3. Mashkina, A. V.; Khairulina, L. N. Reaction of dimethyl disulfide with thiophene catalyzed by zeolite. Russ. J. Org. Chem. 2015, 51 (2), 217-220.