L-Norvaline, 99%
L-Norvaline, 99%
L-Norvaline, 99%
Thermo Scientific Chemicals
L-Norvaline, 99%
CAS: 6600-40-4 | C5H11NO2 | 117.15 g/mol
Have Questions?
Catalog NumberQuantity
ALFL08658.065 g
Catalog number ALFL08658.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialL-Norvaline
Melting Point>300°C
CAS6600-40-4
Recommended StorageAmbient temperatures
EINECS Number229-543-3
View more
L-Norvaline enhances NO production from activated macrophages. It is known to promote tissue regeneration and muscle growth and become a precursor in the penicillin biosynthetic pathway. It also can be used for nutrition agent and drug synthesis. For example: it can react with phthalic acid anhydride to get N,N-phthaloyl-L-norvaline.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
L-Norvaline enhances NO production from activated macrophages. It is known to promote tissue regeneration and muscle growth and become a precursor in the penicillin biosynthetic pathway. It also can be used for nutrition agent and drug synthesis. For example: it can react with phthalic acid anhydride to get N,N-phthaloyl-L-norvaline.
Solubility
Soluble in water (almost transparency), 10.5 g/100 mL (18°C).
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
General References:
  1. Chiung-I Chang, et al. Macrophage Arginase Promotes Tumor Cell Growth and Suppresses Nitric Oxide-mediated Tumor Cytotoxicity.Cancer Res.,2001,611100-1106.
  2. Xiu-Fen Ming, et al. Thrombin Stimulates Human Endothelial Arginase Enzymatic Activity via RhoA/ROCK Pathway Implications for Atherosclerotic Endothelial Dysfunction.Circulation.,2004,1103708-3714.