Trimethyl-1-octylammonium bromide, 97%
Trimethyl-1-octylammonium bromide, 97%
Trimethyl-1-octylammonium bromide, 97%
Trimethyl-1-octylammonium bromide, 97%
Thermo Scientific Chemicals
Trimethyl-1-octylammonium bromide, 97%
CAS: 2083-68-3 | C11H26BrN | 252.24 g/mol
Have Questions?
Catalog NumberQuantity
ALFL08610.065 g
Catalog number ALFL08610.06
Price (MYR)
326.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
326.00
EA
Add to cart
Specifications
Chemical Name or MaterialTrimethyl-1-octylammonium bromide
CAS2083-68-3
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c