N-Phenylthiourea, 97%
N-Phenylthiourea, 97%
N-Phenylthiourea, 97%
N-Phenylthiourea, 97%
Thermo Scientific Chemicals
N-Phenylthiourea, 97%
CAS: 103-85-5 | C7H8N2S | 152.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
10 g
50 g
Catalog number ALFL06690.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialN-Phenylthiourea
CAS103-85-5
Health Hazard 1H300-H317
Health Hazard 2GHS H Statement
H300
Fatal if swallowed.
Health Hazard 3P261-P264b-P270-P272-P280g-P301+P310-P302+P352-P330-P333+P313-P363-P501c
View more
N-phenylthiourea is used in paternity testing prior to the advent of DNA testing. It inhibits melanogenesis and is used to grow transparent fish. It is also used as a repellent for rat, rabbits, weasels and also in medical genetics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-phenylthiourea is used in paternity testing prior to the advent of DNA testing. It inhibits melanogenesis and is used to grow transparent fish. It is also used as a repellent for rat, rabbits, weasels and also in medical genetics.
Solubility
Soluble in hot water and alcohol.
Notes
Incompatible with strong oxidizing agents, strong acids and strong bases.
RUO – Research Use Only
General References:
  1. Mekarat, S.; Pakawatchai, C.; Saithong, S. (N-Phenylthiourea-κS)bis(triphenylphosphane-κP)silver(I) nitrate. Acta. Cryst. 2014, 70 (7), m281-m282.
  2. Farquhar, D. R.; Kovatch, K. J.; Palmer, J. N.; Shofer, F. S.; Adappa, N. D.; Cohen, N. A. Phenylthiocarbamide taste sensitivity is associated with sinonasal symptoms in healthy adults. Int. Forum. Allergy. Rhinol. 2015, 5 (2), 111-118.