1,6-Hexanedithiol, 97%
1,6-Hexanedithiol, 97%
1,6-Hexanedithiol, 97%
1,6-Hexanedithiol, 97%
Thermo Scientific Chemicals
1,6-Hexanedithiol, 97%
CAS: 1191-43-1 | C6H14S2 | 150.30 g/mol
Have Questions?
Catalog NumberQuantity
ALFL06686.1425 g
Catalog number ALFL06686.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material1,6-Hexanedithiol
CAS1191-43-1
Health Hazard 1H227
Health Hazard 2GHS H Statement
H331-H227-H302-H312
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3P210-P280-P370+P378q-P501c
View more
1,6-Hexanedithiol is used in self-assembled monolayers (SAMs). It is used in synthetic rubber and food with spices. It is also used in Food Flavor, Tobacco Flavor, Blending edible flavors: Beef, poultry soup .
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,6-Hexanedithiol is used in self-assembled monolayers (SAMs). It is used in synthetic rubber and food with spices. It is also used in Food Flavor, Tobacco Flavor, Blending edible flavors: Beef, poultry soup .
Solubility
Not miscible in water.
Notes
Incompatible materials are Bases, Oxidizing agents, Reducing agents, Alkali metals. Avoid Heat and direct sunlight.
RUO – Research Use Only
General References:
  1. E. Hutter; J. H. Fendler and D. Roy. Surface Plasmon Resonance Studies of Gold and Silver Nanoparticles Linked to Gold and Silver Substrates by 2-Aminoethanethiol and 1,6-Hexanedithiol.J. Phys. Chem. B.2001,105 (45), 11159-11168.
  2. T. Y. B. Leung; M. C. Gerstenberg; D. J. Lavrich and G. Scoles. 1,6-Hexanedithiol Monolayers on Au(111): A Multitechnique Structural Study.Langmuir.2000,16 (2), 549-561.