4-(Aminomethyl)pyridine, 97+%
4-(Aminomethyl)pyridine, 97+%
4-(Aminomethyl)pyridine, 97+%
4-(Aminomethyl)pyridine, 97+%
Thermo Scientific Chemicals
4-(Aminomethyl)pyridine, 97+%
CAS: 3731-53-1 | C6H8N2 | 108.144 g/mol
Have Questions?
Catalog NumberQuantity
ALFL06345.1425 g
Catalog number ALFL06345.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material4-(Aminomethyl)pyridine
CAS3731-53-1
Health Hazard 1H314-H335
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c