Search Thermo Fisher Scientific
| Catalog Number | Quantity |
|---|---|
| ALFL05936.14 | 25 g |
Suitable for battery materials development.
General Description
• DL-Tryptophan is an α-amino acid, and an essential amino acid precursor of serotonin, melatonin, and vitamin B3
Application
• DL-Tryptophan shows antioxidant activity• It has a role as a Daphnia magna metabolite
• It has been explored in in vitro studies to evaluate the antimicrobial and immunoregulatory functions on indoleamine 2,3-dioxygenase
• It has been used as a medium supplement in Psilocybe atrobrunnea and Psilocybe serbica for the synthesis of compounds
• It can be used as a medium supplement for culturing yeast
General References: