Molecular sieves, 3A, 2-5 mm (0.08-0.20in) beads
Molecular sieves, 3A, 2-5 mm (0.08-0.20in) beads
Molecular sieves, 3A, 2-5 mm (0.08-0.20in) beads
Molecular sieves, 3A, 2-5 mm (0.08-0.20in) beads
Thermo Scientific Chemicals
Molecular sieves, 3A, 2-5 mm (0.08-0.20in) beads
CAS: 308080-99-1 | AlNaO6Si2 | 202.135 g/mol
Have Questions?
Catalog NumberQuantity
ALFL05359.30250 g
Catalog number ALFL05359.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialMolecular sieves, 3A
SensitivityHygroscopic
Quantity250 g
Name Note3 to 5mm (0.12 to 0.20 in) beads
CAS308080-99-1
View more
Molecular sieves, 3A is used as a desiccant in petroleum and chemical industries. It is useful for the drying of unsaturated hydrocarbons such as ethylene, propylene and butadiene. It is also used in the removal of water from ethanol. It finds application in the drying of certain chemicals namely ethanol, refrigerants and natural gas. It plays an important role in selective adsorption of water in insulated glass (IG) and polyurethane. It has been considered as a general-purpose drying agent in polar and non polar media.