Phenyltriethoxysilane, 98%
Phenyltriethoxysilane, 98%
Phenyltriethoxysilane, 98%
Phenyltriethoxysilane, 98%
Thermo Scientific Chemicals
Phenyltriethoxysilane, 98%
CAS: 780-69-8 | C12H20O3Si | 240.37 g/mol
Have Questions?
Catalog NumberQuantity
ALFL04684.0B1000 g
Catalog number ALFL04684.0B
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1000 g
Request bulk or custom format
Specifications
Boiling Point235°C
Chemical Name or MaterialPhenyltriethoxysilane
CAS780-69-8
Health Hazard 1H226-H373
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
Phenyltriethoxysilane is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Phenyltriethoxysilane is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Hydrolyzes in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture. Store under dry inert gas.
RUO – Research Use Only
General References:
  1. M. M. Sprung.; F. O. Guenther. The hydrolysis of n-amyltriethoxysilane and phenyltriethoxysilane. Journal of Polymer Science. 1958, 28 (116), 17-34.
  2. Zhijian Wu.; Hong Xiang.; Taehoon Kim.; Myung-Suk Chun.; Kangtaek Lee. Surface properties of submicrometer silica spheres modified with aminopropyltriethoxysilane and phenyltriethoxysilane. Journal of Colloid and Interface Science. 2006, 304 (1), 119-124.