1-Trimethylsilylmethanol, 95%
1-Trimethylsilylmethanol, 95%
1-Trimethylsilylmethanol, 95%
1-Trimethylsilylmethanol, 95%
Thermo Scientific Chemicals
1-Trimethylsilylmethanol, 95%
CAS: 3219-63-4 | C4H12OSi | 104.224 g/mol
Have Questions?
Catalog NumberQuantity
ALFL04251.031 g
Catalog number ALFL04251.03
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Request bulk or custom format
Specifications
Boiling Point121°C to 122°C
Chemical Name or Material1-Trimethylsilylmethanol
CAS3219-63-4
Health Hazard 1H226
Health Hazard 2GHS H Statement
H226
Flammable liquid and vapor.
View more
1-Trimethylsilylmethanol is used as nucleophile to resolve the racemate obtained from enantioselective esterification of naproxen (2-(6-methoxy-2-naphthyl) propionic acid) by lipases. It was also used as a nucleophilic hydroxymethylation agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Trimethylsilylmethanol is used as nucleophile to resolve the racemate obtained from enantioselective esterification of naproxen (2-(6-methoxy-2-naphthyl) propionic acid) by lipases. It was also used as a nucleophilic hydroxymethylation agent.
Solubility
Fully miscible in water.
Notes
Store at 4°C. Incompatible with oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only