(+/-)-2-Methyl-2,4-pentanediol, 98%
(+/-)-2-Methyl-2,4-pentanediol, 98%
(+/-)-2-Methyl-2,4-pentanediol, 98%
(+/-)-2-Methyl-2,4-pentanediol, 98%
Thermo Scientific Chemicals
(+/-)-2-Methyl-2,4-pentanediol, 98%
CAS: 107-41-5 | C6H14O2 | 118.176 g/mol
Have Questions?
Catalog NumberQuantity
ALFL03187.AE100 mL
Catalog number ALFL03187.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or Material(+/-)-2-Methyl-2,4-pentanediol
CAS107-41-5
Health Hazard 1H227-H315-H319-X
Health Hazard 2GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Health Hazard 3P210-P233-P240-P243-P260-P264b-P271-P280-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P332+P313-P362-P374-P501c
View more
(±)-2-Methyl-2,4-pentanediol is used to control the flow properties of industrial products such as paints, coatings, cleansers, solvents, and hydraulic fluids. It acts as a thickening agent in cosmetic products. It serves as a coupling agent and an additive to hydraulic fluids, inks and cement. It is also employed as a blood volume expander. Further, it is used as a solvent for cleaning and colorant products. In addition to this, it is used in laboratory studies as a precipitant and cryoprotectant in protein crystallography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(^+)-2-Methyl-2,4-pentanediol is used to control the flow properties of industrial products such as paints, coatings, cleansers, solvents, and hydraulic fluids. It acts as a thickening agent in cosmetic products. It serves as a coupling agent and an additive to hydraulic fluids, inks and cement. It is also employed as a blood volume expander. Further, it is used as a solvent for cleaning and colorant products. In addition to this, it is used in laboratory studies as a precipitant and cryoprotectant in protein crystallography.
Solubility
Miscible with water, alcohol, ether, fatty acids and lower aliphatic hydrocarbons. Slightly miscible with carbon tetrachloride.
Notes
Hygroscopic. Incompatible with strong acids, strong oxidizing agents and strong reducing agents.
RUO – Research Use Only
General References:
  1. Reagent for crystallization of proteins.
  2. Grineva, O. V.; Usacheva, T. M.; Zhuravlev, V. I. Crystal conformations of 2-methyl-2,4-pentanediol: Dipole moment calculations and molecular structure optimizations. J. Struct. Chem. 2014, 55 (1), 161-168.
  3. Roussel, G.; Rouse, S. L.; Sansom, M. S. P.; Michaux, C.; Perpète, E. A. The role of 2-methyl-2,4-pentanediol in sodium dodecyl sulfate micelle dissociation unveiled by dynamic light scattering and molecular dynamics simulations. Colloids Surf., B 2014, 114, 357-362.