N,N,N',N'-Tetramethyl-p-phenylenediamine, 98+%
N,N,N',N'-Tetramethyl-p-phenylenediamine, 98+%
N,N,N',N'-Tetramethyl-p-phenylenediamine, 98+%
N,N,N',N'-Tetramethyl-p-phenylenediamine, 98+%
Thermo Scientific Chemicals
N,N,N',N'-Tetramethyl-p-phenylenediamine, 98+%
CAS: 100-22-1 | C10H16N2 | 164.252 g/mol
Have Questions?
Catalog NumberQuantity
ALFL00559.1850 g
Catalog number ALFL00559.18
Price (MYR)
1,243.00
EA
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (MYR)
1,243.00
EA
Add to cart
Specifications
Chemical Name or MaterialN,N,N',N'-Tetramethyl-p-phenylenediamine
CAS100-22-1
Health Hazard 1H302+H312+H332
Health Hazard 2GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
View more
N,N,N',N'-Tetramethyl-p-phenylenediamine is a reducing co-substrate for heme peroxidases. It is used for detection of peroxidases on polyacrylamide gels.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N,N,N′,N′-Tetramethyl-p-phenylenediamine is a reducing co-substrate for heme peroxidases. It is used for detection of peroxidases on polyacrylamide gels.
Solubility
Slightly soluble in cold, more in hot water.
Notes
Air & Light Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
General References:
  1. Powerful electron donor which readily forms stable, dark blue radical cation: J. Am. Chem. Soc., 61, 1981 (1939); 65, 1747 (1943), which is involved in a wide variety of redox reactions:
  2. Electron acceptor in studies of catalytic properties of oxoreductases in cellular organelles: Anal. Biochem., 131, 404 (1983). Reducing agent for cytochrome c.
  3. Compare N,N-Dimethyl-p-phenyl enediamine dihydrochloride, A10343.