Sodium n-dodecyl sulfate, 99%, Molecular Biology Grade
Sodium n-dodecyl sulfate, 99%, Molecular Biology Grade
Sodium n-dodecyl sulfate, 99%, Molecular Biology Grade
Sodium n-dodecyl sulfate, 99%, Molecular Biology Grade
Thermo Scientific Chemicals
Sodium n-dodecyl sulfate, 99%, Molecular Biology Grade
CAS: 151-21-3 | C12H25NaO4S | 288.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
250 g
Catalog number ALFJ67606.36
Price (MYR)
1,698.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
1,698.00
EA
Add to cart
Specifications
Chemical Name or MaterialSodium n-dodecyl sulfate
CAS151-21-3
Health Hazard 1H228-H302+H332-H315-H318-H335-H500
Health Hazard 2GHS H Statement
H228-H311-H302-H315-H319-H335
Flammable solid.
Toxic in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P210-P240-P241-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P370+P378q-P501c