Levofloxacin, 98%
Levofloxacin, 98%
Levofloxacin, 98%
Thermo Scientific Chemicals
Levofloxacin, 98%
Broad-spectrum antibiotic used in pharmacokinetic , antibiotic resistance, and resistance prevention studies. Levofloxacin is active against Gram-positive and Gram-negative bacteria. It inhibits DNA gyrase (type II topoisomerase) and topoisomerase IV, thereby inhibiting cell division. | CAS: 100986-85-4 | C18H20FN3O4 | 361.373 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALFJ66943.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialLevofloxacin
TypeLevofloxacin
Physical FormPowder
CAS100986-85-4
Health Hazard 1H302-H317-H334
View more
Levofloxacin is a broad-spectrum antibiotic used in pharmacokinetic , antibiotic resistance , and resistance prevention studies. Levofloxacin is active against Gram-positive and Gram-negative bacteria. It inhibits DNA gyrase (type II topoisomerase) and topoisomerase IV, thereby inhibiting cell division.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Levofloxacin is a broad-spectrum antibiotic used in pharmacokinetic , antibiotic resistance , and resistance prevention studies. Levofloxacin is active against Gram-positive and Gram-negative bacteria. It inhibits DNA gyrase (type II topoisomerase) and topoisomerase IV, thereby inhibiting cell division.
Solubility
Slightly soluble in water or methanol. Soluble in glacial acetic acid or dichloromethane
Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
RUO – Research Use Only