Picloram, 95%
Picloram, 95%
Picloram, 95%
Thermo Scientific Chemicals
Picloram, 95%
Belongs to the puridine class of the auxinic compounds similar to the hormone Indole-3-acetic acid (IAA) | CAS: 1918-02-1 | C6H3Cl3N2O2 | 241.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFJ66702.14
Price (MYR)
3,978.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
3,978.00
EA
Add to cart
Specifications
Chemical Name or MaterialPicloram
CAS1918-02-1
Health Hazard 1H319
Health Hazard 3P264b-P280i-P305+P351+P338
Melting Point219°C
View more
Belongs to the puridine class of the auxinic compounds similar to the hormone Indole-3-acetic acid (IAA)