Thidiazuron
Thermo Scientific Chemicals
Thidiazuron
220.25 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ66663.MD250 mg
Catalog number ALFJ66663.MD
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mg
Request bulk or custom format
Specifications
Molecular Weight (g/mol)220.25
Quantity250 mg
PackagingPlastic bottle
FormPowder
Unit SizeEA
Thidiazuron is a plant growth regulator used in micropropagations to supplement media such as Murashige and Skoog medium. TDZ promotes plant organogenesis and plant regeneration. Thidiazuron enables the production of virus-free stock plants, speeds up breeding, and provides new genotypes for the market.