Fosfomycin disodium, 90%
Fosfomycin disodium, 90%
Fosfomycin disodium, 90%
Fosfomycin disodium, 90%
Thermo Scientific Chemicals
Fosfomycin disodium, 90%
Prevents peptidoglycan synthesis by inhibiting MurA | CAS: 26016-99-9 | C3H5Na2O4P | 182.02 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ66602.1425 g
Catalog number ALFJ66602.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialFosfomycin disodium
TypeFosfomycin Disodium
Physical FormPowder
CAS26016-99-9
Melting Point>300°C
View more
Prevents peptidoglycan synthesis by inhibiting MurA, an enzyme responsible for synthesizing N-acetylmuramic acid or NAM, a major component of peptidoglycan