Methicillin sodium salt, 90+%
Methicillin sodium salt, 90+%
Methicillin sodium salt, 90+%
Methicillin sodium salt, 90+%
Thermo Scientific Chemicals
Methicillin sodium salt, 90+%
CAS: 132-92-3 | C17H19N2NaO6S | 402.40 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ66414.031 g
Catalog number ALFJ66414.03
Price (MYR)
2,657.00
EA
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
2,657.00
EA
Add to cart
Specifications
Chemical Name or MaterialMethicillin sodium salt
TypeMethicillin Sodium Salt
Physical FormPowder
CAS132-92-3
Health Hazard 1H315-H317-H319-H334-H335
View more
Targets primarily the cell wall of Gram-positive organisms especially Staphylococcus aureus. Methicillin is used to study bacterium susceptibility, and to inhibit cell-wall synthesis. PBP's are enzymes which catalyze a pentaglycine crosslink between alanine and lysine residues providing additional strength to the cell wall. Without a pentaglycine crosslink, the integrity of the cell wall is severely compromised and ultimately leads to cell lysis and death. Methicillin Sodium Salt is inactivated by gastric acid so it is given to cells by other means in which gastric acid is not present in vitro.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Targets primarily the cell wall of Gram-positive organisms especially Staphylococcus aureus. Methicillin is used to study bacterium susceptibility, and to inhibit cell-wall synthesis. PBP′s are enzymes which catalyze a pentagl
RUO – Research Use Only
General References:
  1. Campanella, L.; Mazzei, F.; Sbrilli, R.; Tomassetti, M. Benzylpenicillin PVC membrane electrode for the determination of antibiotics in formulations. Journal of Pharmaceutical and Biomedical Analysis. 1988, 6, (3), 299-305.
  2. Michael Averill Schwartz.; David E. Tutt. Model catalysts which stimulate penicillinase. V. The cycloheptaamylose-catalyzed hydrolysis of penicillins. J. Am. Chem. Soc. 1971, 93, (3), 767-772.