D-Galacturonic acid monohydrate, 97%
D-Galacturonic acid monohydrate, 97%
D-Galacturonic acid monohydrate, 97%
D-Galacturonic acid monohydrate, 97%
Thermo Scientific Chemicals
D-Galacturonic acid monohydrate, 97%
CAS: 91510-62-2 | C6H9O7
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALFJ66282.14
Price (MYR)
903.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
903.00
EA
Add to cart
Specifications
Chemical Name or MaterialD-Galacturonic acid monohydrate
CAS91510-62-2
ColorWhite
Recommended StorageAmbient temperatures
EINECS Number211-682-6
View more
D-(+)-Galacturonic acid monohydrate is a chemical used in the synthesis of N-(D-galacturonoyl) amino acids and dipeptides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
D-(+)-Galacturonic acid monohydrate is a chemical used in the synthesis of N-(D-galacturonoyl) amino acids and dipeptides.
Solubility
Soluble in water
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only
General References:
  1. Chang-Won Cho.; Do-Youn Lee.; Chan-Wha Kim. Concentration and purification of soluble pectin from mandarin peels using crossflow microfiltration system. Carbohydrate Polymers. 2003, 54, (1), 21-26.
  2. Verdini, R.A.; Zorrilla, S.E.; Rubiolo, A.C. Calcium Uptake during Immersion of Strawberries in CaCl2 Solutions. Journal of Food Science. 2008, 73, (7), C533-C539.