Polysorbate 20, Electrophoresis Reagent
Polysorbate 20, Electrophoresis Reagent
Polysorbate 20, Electrophoresis Reagent
Polysorbate 20, Electrophoresis Reagent
Thermo Scientific Chemicals
Polysorbate 20, Electrophoresis Reagent
CAS: 9005-64-5 | (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6 | 522.68 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number ALFJ66278.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialPolysorbate 20
ColorColorless to Yellow
Name NoteElectrophoresis Reagent
CAS9005-64-5
Physical FormLiquid