Sulfadoxine, 98%
Sulfadoxine, 98%
Sulfadoxine, 98%
Thermo Scientific Chemicals
Sulfadoxine, 98%
CAS: 2447-57-6 | C12H14N4O4S | 310.33 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ALFJ66268.1425 gPlastic bottle
Catalog number ALFJ66268.14
Price (MYR)
914.00
EA
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
914.00
EA
Add to cart
Specifications
Chemical Name or MaterialSulfadoxine
CAS2447-57-6
Health Hazard 1H315-H319-H335
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c