Search Thermo Fisher Scientific
| Catalog Number | Quantity |
|---|---|
| ALFJ66237.06 | 5 g |
Suitable for battery materials development.
General Description
• Cefepime hydrochloride is a fourth-generation cephalosporin antibiotic that works against most antibiotic resistant strains of bacteria
• Cefepime hydrochloride is a penicillin binding protein inhibitor that prevents peptidoglycan cross-linkage and leads to cell wall weakening and lysis
Application
• Cefepime hydrochloride monohydrate can be used for the laboratory study of microbial physiology and antibiotic resistance
• Cefepime has broad-spectrum activity; it is appropriate for research on bacteria responsible for resistant infections and physiological studies
General References: