cis-5,8,11,14,17-Eicosapentaenoic acid
cis-5,8,11,14,17-Eicosapentaenoic acid
cis-5,8,11,14,17-Eicosapentaenoic acid
cis-5,8,11,14,17-Eicosapentaenoic acid
Thermo Scientific Chemicals
cis-5,8,11,14,17-Eicosapentaenoic acid
CAS: 10417-94-4 | C20H30O2, C20H30O2 | 302.46 g/mol
Have Questions?
Catalog NumberQuantity
ALFJ66060.MC100 mg
Catalog number ALFJ66060.MC
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mg
Request bulk or custom format
Specifications
Quantity100 mg
DescriptionColorless
Recommended StorageStore at -20°C; Store under Argon
CAS10417-94-4
Molecular FormulaC20H30O2, C20H30O2
View more
Important polyunsaturated fatty acid of the marine food chain that serves as a precursor for the prostaglandin-3 and thromboxane-3 families. It differs from arachidonic acid (the eicosatetraenoic acid that is a precursor for the prostaglandin and thromboxane-2 families) by the extra double bond between the third and fourth carbons from the methyl end of the molecule. Antilipemic.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
-30°C to -10°C